Search Thermo Fisher Scientific
MA1-26836 detects Dystrophin from human, mouse, porcine, rabbit, and rat samples.
MA1-26836 has been successfully used in immunohistochemistry (frozen), and Western blot procedures.
The MA1-26836 immunogen is a recombinant fragment of human Dystrophin.
Dystrophin is the 427kDa protein product of the DMB/BMD gene located on the X chromosome at position Xp21. Western blotting and immunohistochemistry are the two established methods for the detection of abnormalities of dystrophin expression in muscle biopsies. Dystrophin abnormalities are thought to occur in 100% of patients with DMD/BMD, although genetic abnormalities may only be detected in up to 65% of cases.