Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Description: EMAP-II also called SCYE1 is a tumor derived cytokine that plays a role in a wide variety of activities on endothelial cells, monocytes and neutrophils. EMAP-2 is synthesised as a 34 kDa precursor and cleaved enzymatically to produce a biologically active 22 kDa mature polypeptide. The precursor protein is identical to the p43 subunit, which is associated with the multi-tRNA synthetase complex, and it modulates aminoacylation activity of tRNA synthetase in normal cells. EMAP-2 plays a role in in the stimulation of inflammatory responses after proteolytic cleavage in tumor cells. The release of this cytokine renders the tumor-associated vasculature sensitive to tumor necrosis factor.
Applications Tested: ELISA, Western Blotting.
Purity: >97%.
The protein encoded by this gene is a cytokine that is specifically induced by apoptosis, and it is involved in the control of angiogenesis, inflammation, and wound healing. The release of this cytokine renders the tumor-associated vasculature sensitive to tumor necrosis factor. The precursor protein is identical to the p43 subunit, which is associated with the multi-tRNA synthetase complex, and it modulates aminoacylation activity of tRNA synthetase in normal cells. This protein is also involved in the stimulation of inflammatory responses after proteolytic cleavage in tumor cells. Multiple transcript variants encoding different isoforms have been found for this gene. A pseudogene has been identified on chromosome 20.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: aminoacyl tRNA synthase complex-interacting multifunctional protein 1; ARS inducible multifunctional protein 1; endothelial monocyte activating polypeptide 2; mca1; multisynthase complex auxiliary component p43; multisynthetase complex auxiliary component p43; small inducible cytokine subfamily E, member 1
Gene Aliases: 9830137A06Rik; AIMP1/p43; Emap2; EMAPII; p43; Scye1
Entrez Gene ID: (Mouse) 13722
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support