Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: SHQIVPGGRY GVQSFFPEPF VNTLPQARYY NGERTVPQTN GLLSPQQSEE VANPPQRWLV TPVQQPGTNK LDISSYESEY TSSTLLPYGI KSLPLQTSHA LGYYPDPTF
TBR2 is a member of a conserved protein family that shares a common DNA-binding domain, the T-box. T-box genes encode transcription factors involved in the regulation of developmental processes. A similiar protein disrupted in mice is shown to be essential during trophoblast development and gastrulation. TBR2 plays a role in brain development and is required for the specification and the proliferation of the intermediate progenitor cells and their progeny in the cerebral cortex.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: eomesodermin; Eomesodermin homolog; T-box brain protein 2; T-brain-2
Gene Aliases: EOMES; TBR2
UniProt ID: (Human) O95936
Entrez Gene ID: (Human) 8320, (Rat) 316052
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support