Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
OriGene
This Antibody was verified by Knockout to ensure that the antibody binds to the antigen stated.
ERCC1 is a non-catalytic component of a structure-specific DNA repair endonuclease responsible for the 5'-incision during DNA repair. It is responsible, in conjunction with SLX4, for the first step in the repair of interstrand cross-links (ICL). It participates in the processing of anaphase bridge-generating DNA structures, which consist in incompletely processed DNA lesions arising during S or G2 phase, and can result in cytokinesis failure. ERCC1 is also required for homology-directed repair (HDR) of DNA double-strand breaks, in conjunction with SLX4. (Uniprot)
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DNA excision repair protein ERCC-1; excision repair cross-complementation group 1; excision repair cross-complementing rodent repair deficiency, complementation group 1 (includes overlapping antisense sequence)
Gene Aliases: COFS4; ERCC1; RAD10; UV20
UniProt ID: (Human) Q96S40
Entrez Gene ID: (Human) 2067
Molecular Function: endodeoxyribonuclease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support