Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The protein encoded by this gene is a member of the Ser/Thr protein kinase family, and is most closely related to mitogen-activated protein kinases (MAP kinases). MAP kinases also known as extracellular signal-regulated kinases (ERKs), are activated through protein phosphorylation cascades and act as integration points for multiple biochemical signals. This kinase is localized in the nucleus, and has been reported to be activated in fibroblasts upon treatment with serum or phorbol esters. Tissue specificity: Highest expression in the skeletal muscle, followed by the brain. Also found in heart, placenta, lung, liver, pancreas, kidney and skin fibroblasts.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZp686F03189; ERK-3; Extracellular signal-regulated kinase 3; extracellular signal-regulated kinase, p97; MAP kinase 6; MAP kinase isoform p97; MAPK 6; Mitogen-activated protein kinase 6; p97-MAPK; protein kinase, mitogen-activated 5; protein kinase, mitogen-activated 6
Gene Aliases: ERK3; HsT17250; MAPK6; p97MAPK; PRKM6
UniProt ID: (Human) Q16659
Entrez Gene ID: (Human) 5597
Molecular Function: non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support