Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Peptide Sequence: WDPQEVTLQL FTDGITNKLI GCYVGNTMED VVLVRIYGNK TELLVDRDEE VKSFRVLQAH GCAPQLYCTF NNGLCYEFIQ GEALDPKHVC NPAIFR
Ethanolamine kinase 1 (EKI1), functions in the first committed step of the phosphatidylethanolamine synthesis pathway. This cytosolic enzyme is specific for ethanolamine and exhibits negligible kinase activity on choline.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EKI 1; Ethanolamine kinase 1; putative protein product of Nbla10396
Gene Aliases: EKI; EKI 1; EKI1; ETNK1; Nbla10396
UniProt ID: (Human) Q9HBU6
Entrez Gene ID: (Human) 55500
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support