Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This antibody recognizes native and denatured mouse and human nidogen, and blocks binding of nidogen to laminin.
It has been successfully used in ELISA and Western blot applications.
The immunogen is a recombinant fragment corresponding to the G3 region of mouse nidogen produced in Fisher rat plus YB2/0 cells.
Basement membrane components play an important role in cell adhesion, locomotion, differentiation and embryonic compartmentalization. Although the precise composition and structure of basal laminae varies from tissue to tissue, as well as within the same tissue at different developmental stages, most mature basal laminae contain type IV collagen, laminins, heparan sulphate proteoglycans and entactin (or nidogen). Entactin is a sulfated glycoprotein that is widely distributed in basement membranes. It is tightly associated with laminin and also binds to collagen IV. Entactin probably has a role in cell-extracellular matrix interactions.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: enactin; Ent1; Entactin; entactin 1; NID 1; NID-1; Nidogen (enactin); Nidogen precursor (Entactin); Nidogen-1
Gene Aliases: A630025O17; Ent; entactin; entactin-1; NID; NID1; nidogen-1
UniProt ID: (Human) P14543, (Mouse) P10493
Entrez Gene ID: (Human) 4811, (Mouse) 18073
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support