Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Applications | Tested Dilution | Publications |
---|---|---|
Western Blot (WB) |
1:2,000 | - |
Flow Cytometry (Flow) |
0.2 µg | - |
Product Specifications | |
---|---|
Species Reactivity |
Human |
Host/Isotype |
Mouse / IgG1 |
Class |
Monoclonal |
Type |
Antibody |
Clone |
2C5B3 |
Immunogen |
EDN1 Fusion Protein Ag28334 |
Conjugate |
Unconjugated |
Form |
Liquid |
Concentration |
1000 µg/mL |
Storage conditions |
-20°C |
Aliquoting is unnecessary for -20°C storage.
The protein encoded by this gene is proteolytically processed to release a secreted peptide termed endothelin 1. This peptide is a potent vasoconstrictor and is produced by vascular endothelial cells. Endothelin 1 also can affect the central nervous system. Two transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Big endothelin 1; Big endothelin-1; Big ET; Big ET 1; Big ET1; BigET; Endothelin-1; Et; ET-1; PPET1; Preproendothelin-1
Gene Aliases: EDN1
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support