Search Thermo Fisher Scientific
PA1-30380 detects EphA4 from mouse samples.
PA1-30380 has been successfully used in ELISA, Western blot applications.
The PA1-30380 immunogen is: Recombinant fragment (extracellular domain) (Mouse). Expressed in NSO cells.
Ephrin A4 is also known as LERK-4 or ELF-4. Ephrin-A4 binds EphA2, EphA3, EphA4, EphA5, EphA6, EphA7, and EphB1. To date, at least 14 members of the Eph receptor family and a family of 8 ligands have been identified. Ligands of Eph family receptors are structurally related membrane-bound proteins that can be subdivided into two major subclasses: ephrin-A and ephrin-B. Ligands in the ephrin-A subclass, including the prototype family member ephrin-A1 (B61), are membrane associated through glycosylphosphatidyl-inositollinkages; whereas ephrin-B subclass consists of ligands with transmembrane domains.