Search Thermo Fisher Scientific
Mouse anti Estradiol antibody, clone 6E1 recognizes estradiol, a reproductive hormone and the most potent naturally occuring estrogen.
Estradiol (17 beta-estradiol) is a sex hormone. Estradiol is the predominant sex hormone present in females. It is also present in males, being produced as an active metabolic product of testosterone. It represents the major estrogen in humans. Estradiol has not only a critical impact on reproductive and sexual functioning, but also affects other organs including the bones.