Invitrogen
Percent identity with other species by BLAST analysis: Human, Gibbon (100%) Gorilla, Monkey, Marmoset (94%) Bat, Elephant, Horse (88%) Mouse, Bovine, Pig (82%).
Coagulation factor II (thrombin) receptor-like 3 (F2RL3) is a member of the large family of 7-transmembrane-region receptors that couple to guanosine-nucleotide-binding proteins. F2RL3 is also a member of the protease-activated receptor family. F2RL3 is activated by proteolytic cleavage of its extracellular amino terminus. The new amino terminus functions as a tethered ligand and activates the receptor. F2RL3 is activated by thrombin and trypsin.