Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MAFTGKFEME SEKNYDEFMK LLGISSDVIE KAHNFKIVTE VQQDGQDFTW SQHYYGGHTM TNKFTVGKES NIQTMGGKTF KATVQMEGGK LVVNFPNYHQ TSEIVGDKLV EVSTIGGVTY ERVSKRLA
This gene encodes the ileal fatty acid binding protein. Fatty acid binding proteins are a family of small, highly conserved, cytoplasmic proteins that bind long-chain fatty acids and other hydrophobic ligands. FABP6 and FABP1 (the liver fatty acid binding protein) are also able to bind bile acids. It is thought that FABPs roles include fatty acid uptake, transport, and metabolism. Transcript variants generated by alternate transcription promoters and/or alternate splicing have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: fatty acid binding protein 6, ileal; Fatty acid-binding protein 6; Gastrotropin; GT; I-15P; I-BABP; ILBP; ileal bile acid binding protein; Ileal lipid-binding protein; illeal lipid-binding protein; Intestinal 15 kDa protein; Intestinal bile acid-binding protein
Gene Aliases: FABP6; I-15P; I-BABP; I-BALB; I-BAP; ILBP; ILBP3; ILLBP
UniProt ID: (Human) P51161
Entrez Gene ID: (Human) 2172
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support