Search Thermo Fisher Scientific
FIGURE: 1 / 2
Peptide Sequence: MAQEKMELDL ELPPGTGGSP AEGGGSGGGG GLRRSNSAPL IHGLSDTSPV FQAEAPSARR NSTTFPSRHG LLLPASPVRM HSSRLHQIKQ EEGMDLINRE TVHEREVQTA MQISHSWEES FSLSDNDVEK SASPKRIDFI PVSPAPSPTR GIGKQCFSPS LQSFVSSNGL PPSPIPSPTT RFTTRRSQSP INCIRPSVLG PLKRKCEMET EYQPKRFFQG ITNMLSSDVA QLSDPGVCVS SDTLDGNSSS AGSSCNSPAK VSTTTDSPVS PAQAASPFIP LDELSSK
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: PABIR family member 1; PPP2R1A-PPP2R2A-interacting phosphatase regulator 1
Gene Aliases: C9orf42; FAM122A; PABIR1
UniProt ID: (Human) Q96E09
Entrez Gene ID: (Human) 116224
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support