Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MSTNDTIVSG TLPQRMASCL EVLDLFLNQP HMVLSSFVLA EKAAAYRDRD SQRDLVEAVA HILGIRDLAA VNLDSSLADL GLDSLMSVEV RQTLERELNL VLSVREVRQL TLRKLQELSS KADEASELAC PTPKEDGLAQ QQTQLNLRSL LVNPEGPTLM RLNSVQSSER PLFLVHPIEG STTVFHSLAS GLSIPTYGLQ CTRAAPLDSI HSLAAYYIDC IRQVQPEGPY RVAGYSYGAC VAFEMCSQLQ AQQSPAPTHN SLFLFDGSPT YVLAYTQSYR AKLTPGCEAE AETEAICFFV QQFTDMEHNR VLEALLPLKG LEERVAAAVD LIIKSHQGLD RQELSFAARS FYYKLRAAEQ YTPKAKYHGN VMLLRAKTGG AYGEDLGADY NLSQVCDGKV SVHVIEGDHR TLLEGSGLES IISIIHSSLA EPRVSVREG
Fatty acid synthase is a multifunctional protein. Its main function is to catalyze the synthesis of palmitate from acetyl-CoA and malonyl-CoA, in the presence of NADPH, into long-chain saturated fatty acids. In some cancer cell lines, this protein has been found to be fused with estrogen receptor-alpha (ER-alpha), in which the N-terminus of FAS is fused in-frame with the C-terminus of ER-alpha.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Fatty acid synthase; MGC14367; MGC15706; short chain dehydrogenase/reductase family 27X, member 1; Type I fatty acid synthase
Gene Aliases: FAS; FASN; OA-519; SDR27X1
UniProt ID: (Human) P49327
Entrez Gene ID: (Human) 2194
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support