Search Thermo Fisher Scientific
PA5-21620 targets FBL6 in ICC, IF, IHC (P), and WB applications and shows reactivity with Human samples.
The PA5-21620 immunogen is a synthetic peptide corresponding to a region within amino acids 478 and 539 of FBL6 (Uniprot ID#Q8N531).
Substrate-recognition component of the SCF (SKP1-CUL1-F-box protein)-type E3 ubiquitin ligase complex.