Based on 100% sequence identity, this antibody is predicted to react with Mouse and Bovine
Members of the F-box protein family, such as FBXO28, are characterized by an approximately 40-amino acid F-box motif. SCF complexes, formed by SKP1.