Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MEELVVEVR GSNGAFYKAF VKDVHEDSIT VAFENNWQPD RQIPFHDVRF PPPVGYNKDI NESDEVEVYS RANEKEPCCW WLAKVRMIKG EFYVIEYAAC DATYNEIVTI ERLRSVNPNK PATKDTFHKI KLDVPEDLRQ MCAKEAAHKD FKKAVGAFSV TYDPENYQLV ILSINEVTSK RAHMLIDMHF RSLRTKLSLI MRNEEASKQL ESSRQLASRF HEQFIVREDL MGLAIGTHGA NIQQARKVPG VTAIDLDEDT CTFHIYGEDQ DAVKKARSFL EFAEDVIQVP RNLVGLKI (1-297 aa encoded by BC038998)
FMR1 binds RNA and is associated with polysomes. The protein may be involved in mRNA trafficking from the nucleus to the cytoplasm. A trinucleotide repeat in the 5' UTR is normally found at 6-53 copies, but an expansion to 55-230 repeats is the cause of fragile X syndrome. Expansion of the trinucleotide repeat may also cause one form of premature ovarian failure. Multiple alternatively spliced transcript variants that encode different protein isoforms and which are located in different cellular locations have been described for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FMRP; fragile X mental retardation protein 1; fragile X mental retardation protein 1 homolog; fragile X mental retardation syndrome 1 homolog; fragile X mental retardation-1 protein; Fragile X messenger ribonucleoprotein; Fragile X messenger ribonucleoprotein 1; MGC87458; Protein FMR-1; ragile X mental retardation protein
Gene Aliases: Fmr-1; FMR1; FMRP; FRAXA; POF; POF1
UniProt ID: (Human) Q06787, (Rat) Q80WE1
Entrez Gene ID: (Human) 2332, (Rat) 24948, (Mouse) 14265
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support