Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody is tested in Peptide ELISA: antibody detection limit dilution 8,000.
This gene belongs to the forkhead family of transcription factors which is characterized by a distinct forkhead domain. The specific function of this gene has not yet been determined; however, it is possible that this gene plays an important role in the development of the cochlea and vestibulum, as well as embryogenesis. Mutations in this gene may be associated with the common cavity phenotype. Two transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Forkhead; Forkhead box protein I1; forkhead-like 10; forkhead-related activator 6; Forkhead-related protein FKHL10; Forkhead-related transcription factor 6; FREAC-6; Hepatocyte nuclear factor 3 forkhead homolog 3; HFH-3; HNF-3/fork-head homolog 3; HNF-3/fork-head homolog-3; MGC34197
Gene Aliases: FKH10; FKHL10; FOXI1; FREAC-6; FREAC6; HFH-3; HFH3
UniProt ID: (Human) Q12951
Entrez Gene ID: (Human) 2299
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support