Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MDIIKGNLDG ISKPASNSRI RPGSRSSNAS LEVLSTEPGS FKVDTASNLN SGKEDHSESS NTENRRTSND DKQESCSEKI KLAEEGSDED LDLVQHQIIS ECSDEPKLKE LDSQLQDAIQ KMKKLDKILA KKQRREKEIK KQGLEMRIKL WEEIKSAKYS EAWQSKEEME NTKKFLSLTA VSEETVGPSH EEEDTFSSVF HTQIPPEEYE MQMQKLNKDF TCDVERNESL IKSGKKPFSN TEKIELRGKH NQDFIKRNIE LAKESRNPVV MVDREKKRLV ELLKDLDEKD SGLSSSEGDQ SGWVVPVKGY ELAVTQHQQL AEIDIKLQEL SAASPTISSF SPRLENRNNQ KPDHDGERNM EVTPGEKILR NTKEQRDLHN RLREIDEKLK MMKENVLEST SRLSEEQLKC LLDECILKQK SIIKLSSERK KEDIEDVTPV FPQLSRSIIS KLLNESETKV QKTEVEDADM LESEECEASK GYYLTKALTG HNMSEALVTE AENMKCLQFS KDVIISDTKD YFMSKTLGIG RLKRPSFLDD PLYGISVSLS SEDQHLKLSS PENTIADEQE TKDAAEECKE P
The exact functions of FSIP1 remain unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Fibrous sheath-interacting protein 1
Gene Aliases: FSIP1; HSD10
UniProt ID: (Human) Q8NA03
Entrez Gene ID: (Human) 161835
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support