The recommended shelf life for this product is 1 year from date of receipt.
Based on 100% sequence identity, this antibody is predicted to react with Orangutan
Although the function of this gene is not known, the existence of this gene is supported by mRNA and EST data. A possible function of the encoded protein can be inferred from amino acid sequence similarity to the E.coli FtsJ protein and to a mouse protein possibly involved in embryogenesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 2'-O-ribose RNA methyltransferase SPB1 homolog; ectoplacental cone, invasive trophoblast giant cells, extraembryonic ectoderm and chorion sequence 3; FLJ20062; FTSJ3; pre-rRNA 2'-O-ribose RNA methyltransferase FTSJ3; pre-rRNA processing protein FTSJ3; Protein ftsJ homolog 3; Putative rRNA methyltransferase 3; rRNA (uridine-2'-O-)-methyltransferase 3; SPB1 RNA methyltransferase homolog
Gene Aliases: AA537063; AU045295; C79843; D11Ertd400e; EPCS3; FTSJ3; SB92; SPB1
UniProt ID: (Human) Q8IY81, (Mouse) Q9DBE9
Entrez Gene ID: (Human) 117246, (Mouse) 56095
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support