Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: SEMVPGAADC NITADSSVYP QADAVIVHHW DIMYNPSANL PPPTRPQGQR WIWFSMESPS NCRHLEALDG
Catalyzes preferentially the transfer of L-fucose, from a guanosine diphosphate-beta-L-fucose, to the N-acetyl-beta-D-glucosamine (GlcNAc) of an N-acetyllactosamine unit (type 2 chain) of an oligosacccharide, or a glycoprotein- and a glycolipid-linked N-acetyllactosamine unit via an alpha (1,3) linkage and participates to the surface espression of VIM-2, Lewis X/SSEA-1 and sialyl Lewis X antigens (PubMed:14718375, PubMed:1740457, PubMed:7721776, PubMed:9737988, PubMed:17604274, PubMed:9737989). Preferentially transfers fucose to the GlcNAc of an internal N-acetyllactosamine unit of a poly-N-acetyllactosamine chain acceptor substrate (PubMed:7721776, PubMed:17604274). Also catalyzes to a lesser extend the transfer of L-fucose to the GlcNAc of a type 1 (beta-D-galactosyl-(1->3)-N-acetyl-beta-D-glucosaminyl) or H-type 1 (alpha-L-Fuc-(1->2)-beta-D-Gal-(1->3)-D-GlcNAc) chain oligosacccharide via an alpha (1,4) linkage (PubMed:14718375, PubMed:1740457, PubMed:7721776, PubMed:9737988, PubMed:17604274). Preferentially catalyzes sialylated type 2 oligosacccharide acceptors over neutral type 2 or H type 2 (alpha-L-Fuc-(1->2)-beta-D-Gal-(1->4)-D-GlcNAc) oligosacccharide acceptors (PubMed:1740457, PubMed:9737989). Lactose-based structures are also acceptor substrates (PubMed:1740457, PubMed:7721776). [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 3-galactosyl-N-acetylglucosaminide 4-alpha-L-fucosyltransferase FUT5; 4-galactosyl-N-acetylglucosaminide 3-alpha-L-fucosyltransferase FUT5; alpha (1,3) fucosyltransferase; Fuc-TV; Fucosyltransferase 5; fucosyltransferase 5 (alpha (1,3) fucosyltransferase); Fucosyltransferase V; fucT-V; Galactoside 3-L-fucosyltransferase
Gene Aliases: FUC-TV; FUT5
UniProt ID: (Human) Q11128
Entrez Gene ID: (Human) 2527
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support