Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
MA1-43005 detects Factor V in human samples.
MA1-43005 has successfully been used in Western blot and ELISA applications.
MA1-43005 was derived from human factor V and the heavy chain (fragment D, 94,000) of factor Va.
Coagulation factor V (or Proaccelerin) is an essential factor of the blood coagulation cascade. This factor circulates in plasma, and is converted to the active form by the release of the activation peptide by thrombin during coagulation. This generates a heavy chain and a light chain which are held together by calcium ions. The active factor V is a cofactor that participates with activated coagulation factor X to activate prothrombin to thrombin. Defects in the Factor V gene can result various diseases including Factor V deficiency, thrombophilia due to activated protein C resistance, Budd-Chiari syndrome, Ischemic stroke, and recurrent pregnancy loss.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Activated protein C cofactor; Coagulation factor V; coagulation factor V (proaccelerin, labile factor); coagulation factor V jinjiang A2 domain; Factor 5; factor V Leiden; Factor5; Proaccelerin, labile factor
Gene Aliases: F5; FVL; PCCF; RPRGL1; THPH2
UniProt ID: (Human) Q8WWQ6
Entrez Gene ID: (Human) 2153
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support