Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MPAQPAPPK PQPTTRTPPQ SQTPGALPAK REQPPSLTRN GPLSCGQRLR KSLSSMTRVV GGLVALRGAH PYIAALYWGH SFCAGSLIAP CWVLTAAHCL QDRPAPEDLT VVLGQERRNH SCEPCQTLAV RSYRLHEAFS PVSYQHDLAL LRLQEDADGS CALLSPYVQP VCLPSGAARP SETTLCQVAG WGHQFEGAEE YASFLQEAQV PFLSLERCSA PDVHGSSILP GMLCAGFLEG GTDACQGDSG GPLVCEDQAA ERRLTLQGII SWGSGCGDRN KPGVYTDVAY YLAWIREHTV S (316-615 aa encoded by BC012390)
This single chain zymogen is converted to a two-chain serine protease with an heavy chain (alpha-factor XIIa) and a light chain. The heavy chain contains two fibronectin-type domains, two epidermal growth factor (EGF)-like domains, a kringle domain and a proline-rich domain, whereas the light chain contains only a catalytic domain. On activation, further cleavages takes place in the heavy chain, resulting in the production of beta-factor XIIa light chain and the alpha-factor XIIa light chain becomes beta-factor XIIa heavy chain. Prekallikrein is cleaved by factor XII to form kallikrein, which then cleaves factor XII first to alpha-factor XIIa and then to beta-factor XIIa. The active factor XIIa participates in the initiation of blood coagulation, fibrinolysis, and the generation of bradykinin and angiotensin. It activates coagulation factors VII and XI. Defects in this gene do not cause any clinical symptoms and the sole effect is that whole-blood clotting time is prolonged.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: beta-factor XIIa part 1; beta-factor XIIa part 2; Coagulation factor XII; coagulation factor XII (Hageman factor); coagulation factor XIIa heavy chain; coagulation factor XIIa light chain; F13; Factor 12; Factor12; HAF; Hageman factor
Gene Aliases: F12; FXII; HAE3; HAEX; HAF
UniProt ID: (Human) P00748, (Mouse) Q80YC5
Entrez Gene ID: (Human) 2161, (Mouse) 58992
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support