Product References
Bovine neutrophil chemotaxis to Listeria monocytogenes in neurolisteriosis depends on microglia-released rather than bacterial factors.
Journal of neuroinflammation
Bagatella S,Haghayegh Jahromi N,Monney C,Polidori M,Gall FM,Marchionatti E,Serra F,Riedl R,Engelhardt B,Oevermann A
MA5-11751 was used in Immunohistochemistry (Paraffin), Western Blot to provide evidence that host-derived rather than bacterial factors trigger PMN chemotaxis to bacterial foci in the CNS, that microglia have a primary role as initiators of bovine PMN chemotaxis into the brain during neurolisteriosis and that blockade of these factors could be a therapeutic target to limit intrathecal PMN chemotaxis and PMN associated damage in neurolisteriosis.
Fri Dec 16 00:00:00 EST 2022
Transcriptome of microglia reveals a species-specific expression profile in bovines with conserved and new signature genes.
Glia
Tavares-Gomes L,Monney C,Neuhaus G,Francisco D,Solis D,Summerfield A,Erny D,Jagannathan V,Oevermann A
Published figure using Factor XIII A monoclonal antibody (Product # MA5-11751) in Immunohistochemistry
Sun Aug 01 00:00:00 EDT 2021
Transcriptome of microglia reveals a species-specific expression profile in bovines with conserved and new signature genes.
Glia
Tavares-Gomes L,Monney C,Neuhaus G,Francisco D,Solis D,Summerfield A,Erny D,Jagannathan V,Oevermann A
Published figure using Factor XIII A monoclonal antibody (Product # MA5-11751) in Immunohistochemistry
Sun Aug 01 00:00:00 EDT 2021
Malignant dermatofibroma: clinicopathological, immunohistochemical, and molecular analysis of seven cases.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc
Mentzel T,Wiesner T,Cerroni L,Hantschke M,Kutzner H,Rütten A,Häberle M,Bisceglia M,Chibon F,Coindre JM
MA5-11751 was used in immunohistochemistry to report on seven cases of malignant dermatofibroma
Fri Feb 01 00:00:00 EST 2013
Procoagulant platelets form an α-granule protein-covered "cap" on their surface that promotes their attachment to aggregates.
The Journal of biological chemistry
Abaeva AA,Canault M,Kotova YN,Obydennyy SI,Yakimenko AO,Podoplelova NA,Kolyadko VN,Chambost H,Mazurov AV,Ataullakhanov FI,Nurden AT,Alessi MC,Panteleev MA
MA5-11751 was used in western blot to study the role of the alpha-granule-containing cap structure of pro-coagulant platelets in their ability to attach to aggregates
Fri Oct 11 00:00:00 EDT 2013
Plaque-like CD34-positive dermal fibroma ("medallion-like dermal dendrocyte hamartoma"): clinicopathologic, immunohistochemical, and molecular analysis of 5 cases emphasizing its distinction from superficial, plaque-like dermatofibrosarcoma protuberans.
The American journal of surgical pathology
Kutzner H,Mentzel T,Palmedo G,Hantschke M,Rütten A,Paredes BE,Schärer L,Guillen CS,Requena L
MA5-11751 was used in immunohistochemistry to characterize the plaque-like CD34-positive dermal fibroma
Mon Feb 01 00:00:00 EST 2010
Transglutaminase 2 is a marker of chondrocyte hypertrophy and osteoarthritis severity in the Hartley guinea pig model of knee OA.
Osteoarthritis and cartilage
Huebner JL,Johnson KA,Kraus VB,Terkeltaub RA
MA5-11751 was used in immunohistochemistry to study transglutaminase-2 as a marker of chondrocyte hypertrophy and disease severity in a model of osteoarthritis
Sat Aug 01 00:00:00 EDT 2009
Dermatomyofibroma: clinicopathologic and immunohistochemical analysis of 56 cases and reappraisal of a rare and distinct cutaneous neoplasm.
The American Journal of dermatopathology
Mentzel T,Kutzner H
MA5-11751 was used in immunohistochemistry to investigate clinicopathologic spectrum of dermatomyofibroma
Sun Feb 01 00:00:00 EST 2009
Molecular MRI of early thrombus formation using a bimodal alpha2-antiplasmin-based contrast agent.
JACC. Cardiovascular imaging
Miserus RJ,Herías MV,Prinzen L,Lobbes MB,Van Suylen RJ,Dirksen A,Hackeng TM,Heemskerk JW,van Engelshoven JM,Daemen MJ,van Zandvoort MA,Heeneman S,Kooi ME
MA5-11751 was used in immunohistochemistry to investigate the effectiveness of a novel MRI contrast agent for the analysis of early thrombus formation
Sat Aug 01 00:00:00 EDT 2009
Normal human dermis contains distinct populations of CD11c+BDCA-1+ dendritic cells and CD163+FXIIIA+ macrophages.
The Journal of clinical investigation
Zaba LC,Fuentes-Duculan J,Steinman RM,Krueger JG,Lowes MA
MA5-11751 was used in immunohistochemistry to study CD11c+BDCA-1+ dendritic cell and CD163+FXIIIA+ macrophage populations in human dermis
Sat Sep 01 00:00:00 EDT 2007
The receptor for advanced glycation end products is highly expressed in the skin and upregulated by advanced glycation end products and tumor necrosis factor-alpha.
The Journal of investigative dermatology
Lohwasser C,Neureiter D,Weigle B,Kirchner T,Schuppan D
MA5-11751 was used in immunohistochemistry to investigate the expression of receptor for advanced glycation end products and its regulatary mechanism in skin
Wed Feb 01 00:00:00 EST 2006
Incidental calcifying fibrous tumor of the stomach presenting as a polyp.
Pathology international
Elpek GO,Küpesiz GY,Ogüs M
MA5-11751 was used in immunohistochemistry to report on a case of incidental calcifying fibrous tumor of the stomach presenting as a polyp
Sat Apr 01 00:00:00 EST 2006