Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Positive Control: PC-3M, rat brain tissue, human lung cancer tissue, human liver tissue, human spleen tissue, HUVEC.
Caveolae are small domains on the inner cell membrane involved in vesicular trafficking and signal transduction. FLOT1 encodes a caveolae-associated, integral membrane protein. The function of flotillin 1 has not been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DADB-118P11.2; Flotillin-1; integral membrane component of caveolae; OTTHUMP00000164855
Gene Aliases: FLOT1
UniProt ID: (Human) O75955
Entrez Gene ID: (Human) 10211
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support