Search Thermo Fisher Scientific
The MA1-7387 antibody reacts with Francisella tularensis LPS in bacterial and infected tissue samples.
MA1-7387 has been successfully used in ELISA and Immunofluorescence applications.
The MA1-7387 antibody was raised against Francisella Lipopolysaccharide.
Francisella tularensis is a small, nonmotile, aerobic, gram-negative coccobacillus. It has a thin lipopolysaccharide-containing envelope and is a hardy non-spore-forming organism that survives for weeks at low temperatures in water, moist soil, hay, straw, and decaying animal carcasses. Lipopolysaccaride (LPS) is a main species-specific antigen of Francisella tularensis. According to the data available LPS of tularemia microbe differs from LPS of other gram-negative bacteria. LPS of tularemia microbe hasn't yet revealed the properties of classical endotoxin, which may be connected with non-typical structure of lipid A, a toxic component of LPS.