Search Thermo Fisher Scientific
PA1-32666 detects GPCR GPR57 from human samples.
PA1-32666 has been successfully used in immunohistochemistry (paraffin tissue) applications.
The PA1-32666 immunogen is: Synthetic peptide, KLH conjugated.
Olfactory receptor activated by several primary trace amines, including isoamylamine. Activated by isoamylamine and cyclohexylamine, but not to the corresponding alcohols, isoamylalcohol and cyclohexanol. This receptor is probably mediated by the G(s)-class of G-proteins which activate adenylate cyclase. [GeneCards]