Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAEQVALSR TQVCGILREE LFQGDAFHQS DTHIFIIMGA SGDLAKKKIY PTIWWLFRDG LLPENTFIVG YARSRLTVAD IRKQSEPFFK ATPEEKLKLE DFFARNSYVA GQYDDAASYQ RLNSHMNALH LGSQANRLFY LALPPTVYEA VTKNIHESCM SQIGWNRIIV EKPFGRDLQS SDRLSNHISS LFREDQIYRI DHYLGKEMVQ NLMVLRFANR IFGPIWNRDN IACVILTFKE PFGTEGRGGY FDEFGIIRDV MQNHLLQMLC LVAMEKPAST NSDDVRDEKV KVLKCISEVQ ANNVVLGQYV GNPDGEGEAT KGYLDDPTVP RGSTTATFAA VVLYVENERW DGVPFILRCG KALNERKAEV RLQFHDVAGD IFHQQCKRNE LVIRVQPNEA VYTKMMTKKP GMFFNPEESE LDLTYGNRYK NVKLPDAYER LILDVFCGSQ MHFVRSDELR EAWRIFTPLL HQIELEKPKP IPYIYGSRGP TEADELMKRV GFQYEGTYKW VNPHKL (1-515 aa encoded by B C000337)
This gene encodes glucose-6-phosphate dehydrogenase. This protein is a cytosolic enzyme encoded by a housekeeping X-linked gene whose main function is to produce NADPH, a key electron donor in the defense against oxidizing agents and in reductive biosynthetic reactions. G6PD is remarkable for its genetic diversity. Many variants of G6PD, mostly produced from missense mutations, have been described with wide ranging levels of enzyme activity and associated clinical symptoms. G6PD deficiency may cause neonatal jaundice, acute hemolysis, or severe chronic non-spherocytic hemolytic anemia. Two transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: G6PD; Glucose-6-phosphate 1-dehydrogenase; Glucose-6-phosphate 1-dehydrogenase X; glucose-6-phosphate dehydrogenase X-linked
Gene Aliases: G28A; G6PD; G6pd-1; G6PD1; G6pdx; Gpdx
UniProt ID: (Human) P11413, (Rat) P05370, (Mouse) Q00612
Entrez Gene ID: (Human) 2539, (Rat) 24377, (Mouse) 14381
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support