Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Marmoset (100%) Rabbit (80%).
Receptor for lysophosphatidic acid (LPA), a mediator of diverse cellular activities.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: G protein-coupled receptor 92; G-protein coupled receptor 92; G-protein coupled receptor 93; LPA receptor 5; LPA-5; Lysophosphatidic acid receptor 5
Gene Aliases: GPR92; GPR93; KPG_010; LPA5; LPAR5
UniProt ID: (Human) Q9H1C0
Entrez Gene ID: (Human) 57121
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support