Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: QAEPKAAEQL ATTTCHALPP AALLCGMREM REVGAGGCCH ALPATGILAF PKLVASVSES GLQAQHGVKI HCRL
Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 68%.
May be involved in neurite outgrowth. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support