Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: CATRPELSRC VAFLDGVAEY EVTPDDKRKA CGRQLTQNFL SHTGPDLIPE VPRQLVTNCT QRLEQGPCKD LFQELTRLTH EYLSVAPFAD YLDSIYFNRF
GRK6 is a member of the guanine nucleotide-binding protein coupled receptor kinase subfamily of the serine/threonine protein kinase family. GRK6 phosphorylates the activated forms of G protein-coupled receptors to initiate their deactivation. Several transcript variants encoding different isoforms have been described for GRK6.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EC 2.7.11.16; G protein-coupled receptor kinase 6; G protein-coupled receptor kinase GRK6; kinase GRK6
Gene Aliases: GPRK6; GRK6
UniProt ID: (Human) P43250
Entrez Gene ID: (Human) 2870
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support