Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Reconstitution: 1 X 50 µL double distilled water (DDW), depending on the sample size. The antibody ships as a lyophilized powder at room temperature. Upon arrival, it should be stored at -20C. The reconstituted solution can be stored at 4C for up to 1 week. For longer periods, small aliquots should be stored at -20C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min).
The GLRA3 gene encodes the alpha-3 subunit of the neuronal glycine receptor, a ligand-gated chloride channel composed of ligand-binding alpha and structural beta polypeptides.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: glycine receptor; Glycine receptor subunit alpha-3; glycine receptor, alpha 3; glycine receptor, alpha 3 subunit; glycine receptor, alpha-3 polypeptide; ligand gated ion channel
Gene Aliases: GLRA3
UniProt ID: (Human) O75311, (Mouse) Q91XP5
Entrez Gene ID: (Human) 8001, (Rat) 114516, (Mouse) 110304
Molecular Function: ligand-gated ion channel
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support