Invitrogen
This Antibody was verified by Relative expression to ensure that the antibody binds to the antigen stated.
FIGURE: 1 / 9
MA1-22144 detects golgi protein, 58K in human, rat, canine, and hamster samples. This antibody is expected to cross react with porcine and bovine Golgi protein due to sequence homology.
MA1-22144 has been successfully used in immunocytochemistry, immunofluorescence, immunohistochemistry (paraffin), immunomicroscopy and Western blot procedures.
The MA1-22144 immunogen is purified, full length, native, rat Golgi protein from rat liver.
Store at 4ºC for short term. For extended storage, aliquot and store at -20ºC, Avoid Freeze/Thaw Cycles.
Golgi Protein, 58K is a bifunctional enzyme that channels 1-carbon units from formiminoglutamate, a metabolite of the histidine degradation pathway, to the folate pool. Mutations in this gene are associated with glutamate formiminotransferase deficiency. Alternatively spliced transcript variants have been found for this gene.