Search Thermo Fisher Scientific
MA1-25751 detects Granulocyte from human samples. MA1-25751 will not react with rat.
MA1-25751 has been successfully used in immunohistochemistry (frozen), and immunohistochemistry (paraffin) procedures.
The MA1-25751 immunogen is nuclei from pokeweed mitogen-stimulated human peripheral blood lymphocytes.
Granulocytes are a category of white blood cells characterized by the presence of granules in their cytoplasm. They are also called polymorphonuclear leukocytes (PMN or PML) because of the varying shapes of the nucleus, which is usually lobed into three segments. In common parlance, the term polymorphonuclear leukocyte often refers specifically to neutrophil granulocytes, the most abundant of the granulocytes. Granulocytes or PMN are released from the bone marrow by the regulatory complement proteins.