Invitrogen
MA5-18004 targets H-2KdDd in FACS and IHC (F) applications and shows reactivity with Mouse samples.
The MA5-18004 immunogen is mouse H-2KdDd.
This protein is an MHC class I alloantigen, which is involved in antigen presentation to T cells expressing CD3/TCR and CD8.
HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.