Invitrogen
HAV 001-02-02 detects Boutulinum toxoid. This antibody does not cross react with type A or type F toxoids.
HAV 001-02-02 has been successfully used in ELISA procedures. HAV 001-02 reacts with botulinum toxoid type E when tested in direct ELISA with type E toxoid coated directly onto the microtiter well. No or very little reaction was seen with toxoids type A and type F.
The HAV 001-02-02 immunogen is Clostridium botulinum type E toxoid.
Epitope of HAV 001-02-02 differs from that of HAV 001-06-02.
NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/mL
Clostridium botulinum is a gram-positive, anaerobic motile bacteria normally found in soil. C. botulinum is a potential threat to both humans and animals through contaminated food because of the potent neurotoxin they produce. C. botulinum bacteria are categorized within 7 strains A-G based on the characteristics of their toxin. A,B, E and F strains are known to cause food poisoning in humans.