Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MGCMKSKFLQ VGGNTSSKTE TSASPHCPVY VPDPTSTIKP GPNSHNSNTP GIREAGSEDI IVVALYDYEA IHHEDLSFQK GDQMVVLEES GEWWKARSLA TRKEGYIPSN YVARVDSLET EEWFFKGISR KDAERQLLAP GNMLGSFMIR DSETTKGSYS LSVRDYDPRQ GDTVKHYKIR TLDNGGFYIS PRSTFSTLQE LVDHYKKGND GLCQKLSVPC MSSKPQKPWE KDAWEIPRES LKLEKKLGAG QFGEVWMATY NKHTKVAVKT MKPGSMSVEA FLAEANVMKT LQHDKLVKLH AVVTKEPIYI ITEFMAKGSL LDFLKSDEGS KQPLPKLIDF SAQIAEGMAF IEQRNYIHRD LRAANILVSA SLVCKIADFG LARVIEDNEY TAREGAKFPI KWTAPEAINF GSFTIKSDVW SFGILLMEIV TYGRIPYPGV SNPEVIRALE RGYRMPRPEN CPEELYNIMM RCWKNRPEER PTFEYIQSVL DDFYTATESQ YQQQP
Hematopoietic cell kinase (HCK) is a SRC-related kinase expressed in lymphoid and hematopoietic cells and may play a role in some forms of leukemia. (HCK Alias: DKFZp686D0638). HCK is a protein-tyrosine kinase that is predominantly expressed in hemopoietic cell types. The encoded protein may help couple the Fc receptor to the activation of the respiratory burst. In addition, it may play a role in neutrophil migration and in the degranulation of neutrophils. Alternate translation initiation site usage, including a non-AUG (CUG) codon, results in the production of two different isoforms, that have different subcellular localization.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Hematopoietic cell kinase; Hemopoietic cell kinase; p59-HCK/p60-HCK; p59Hck; p61Hck; Tyrosine-protein kinase HCK
Gene Aliases: HCK; JTK9; p59Hck; p61Hck
UniProt ID: (Human) P08631
Entrez Gene ID: (Human) 3055
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support