Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 6
Store vial at -20° C prior to opening. Aliquot contents and freeze at -20° C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4° C as an undiluted liquid. Dilute only prior to immediate use.
Anti-HDAC-1 antibody is directed against human HDAC-1 protein. A BLAST analysis was used to suggest reactivity with this protein from human, mouse, rat and chimpanzee sources based on 100% homology for the immunogen sequence. Cross reactivity may occur with HDAC-1 from bovine (82% homology) and chicken (80% homology) sources. Cross reactivity with HDAC-1 homologues from other sources has not been determined.
Histone acetylation and deacetylation, catalyzed by multisubunit complexes, play a key role in the regulation of eukaryotic gene expression. HDAC1 belongs to the histone deacetylase/acuc/apha family and is a component of the histone deacetylase complex. It also interacts with retinoblastoma tumor-suppressor protein and this complex is a key element in the control of cell proliferation and differentiation. Together with metastasis-associated protein-2, HDAC1 deacetylates p53 and modulates its effect on cell growth and apoptosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZp686H12203; HD1; Histone deacetylase 1; Protein deacetylase HDAC1; Protein decrotonylase HDAC1; reduced potassium dependency, yeast homolog-like 1; RP4-811H24.2
Gene Aliases: GON-10; HD1; HDAC1; RPD3; RPD3L1
UniProt ID: (Human) Q13547
Entrez Gene ID: (Human) 3065
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support