Search Thermo Fisher Scientific
FIGURE: 1 / 1
A suggested positive control for this product is BeWo cells.
This protein belongs to the basic helix-loop-helix family of transcription factors. It is a transcriptional repressor of genes that require a bHLH protein for their transcription. The protein has a particular type of basic domain that contains a helix interrupting protein that binds to the N-box rather than the canonical E-box.