Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
High mobility group (HMG) proteins are nonhistone chromosomal proteins.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: High mobility group protein 2-like 1; high-mobility group protein 2-like 1; HMG box domain containing 4; HMG box-containing protein 4; HMG domain-containing protein 4; Protein HMGBCG; Protein THC211630
Gene Aliases: HMG2L1; HMGBCG; HMGXB4; THC211630
UniProt ID: (Human) Q9UGU5
Entrez Gene ID: (Human) 10042
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support