Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide Sequence: GYDALRPFPA SYGASSLPDK TYTSPCFYQQ SNSVLACNRA SYEYGASCFY SDKDLSGASP SGSGKQRGPG DYLHFSPEQQ YKPDSSSGQG KALHDEGADR
In vertebrates, the genes encoding the class of transcription factors called homeobox genes are found in clusters named A, B, C, and D on four separate chromosomes. Expression of these proteins is spatially and temporally regulated during embryonic development. This gene is part of the A cluster on chromosome 7 and encodes a DNA-binding transcription factor which may regulate gene expression, morphogenesis, and differentiation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: homeo box 1B; homeo box A6; Homeobox protein Hox-1B; Homeobox protein Hox-A6; homeobox protein HOXA6
Gene Aliases: HOX1; HOX1.2; HOX1B; HOXA6
UniProt ID: (Human) P31267
Entrez Gene ID: (Human) 3203
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support