Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MDYNRMNSFL EYPLCNRGPS AYSAHSAPTS FPPSSAQAVD SYASEGRYGG GLSSPAFQQN SGYPAQQPPS TLGV
Sequence-specific transcription factor which is part of a developmental regulatory system that provides cells with specific positional identities on the anterior-posterior axis. Acts on the anterior body structures.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Homeobox protein Hox-2I; Homeobox protein Hox-B1; Hox-29
Gene Aliases: HCFP3; Hox-2.9; HOX2; HOX2I; HOXB1
UniProt ID: (Human) P14653
Entrez Gene ID: (Human) 3211
Molecular Function: homeodomain transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support