Search Thermo Fisher Scientific
MA1-7054 detects the human Pampilloma virus.
MA1-7054 has been successfully used in ELISA and Western blot procedures.
The MA1-7054 immunogen is purified recombinant E7 protein of HPV types 16 and 18.
Papillomaviruses are a diverse group of DNA-based viruses that infect the skin and mucous membranes of humans and a variety of animals. 100 different HPV types have been identified to date. Various types of HPV may cause warts while others may cause a subclinical infection resulting in precancerous lesions. All HPVs are transmitted by skin-to-skin contact.