Invitrogen
FIGURE: 1 / 1
Purity is ~98% by SDS PAGE.
Mouse anti Human Heparanase-1 antibody, clone HP3/17 recognizes heparanase-1, also know as HPA1, an endo-beta-D-glucuronidase that degrades heparan sulfate proteoglycans (HSPGs) in the extracellular matrix through cleavage of the heparan sulfate side chains.
Stem cells offer a therapeutic promise for many diseases. In the case of diabetes, stem cells offer the promise that beta cells may be generated ex vivo for the possible transplantation and cure of Type I diabetes (Couri CE, et al. ). For this to happen, stem cells will need to be isolated and expanded, they will need to be differentiated toward the beta cell lineage, and the differentiated cell progeny used for transplantation may need to be isolated from complex cell mixtures following differentiation. Appropriate markers targeting cell surface molecules on developing and mature pancreatic cells will be required for such research.