Search Thermo Fisher Scientific
This antibody is predicted to react with non-human primate based on sequence homology.
The enzyme 17-beta hydroxysteroid dehydrogenase-12 (HSD17B12) uses NADPH to reduce 3-ketoacyl-CoA to 3-hydroxyacyl-CoA during the second step of fatty acid elongation.