Invitrogen
PA1-27202 detects Hepatitis C Virus from virus samples.
PA1-27202 has been successfully used in Western blot and ELISA procedures. By Western blot, a band of 56 kDa is predicted to be observed.
The PA1-27202 immunogen is recombinant, full length core protein of genotype 1a.
HCV is a positive, single-stranded RNA virus in the Flaviviridae family. The genome is approximately 10,000 nucleotides and encodes a single polyprotein of about 3,000 amino acids. The polyprotein is processed by host cell and viral proteases into three major structural proteins and several non-structural protein necessary for viral replication. Several different genotypes of HCV with slightly different genomic sequences have since been identified that correlate with differences in response to treatment with interferon alpha.