Invitrogen
The MA1-7265 antibody reacts with Herpes simplex virus 1&2 Env in viral and infected tissue samples.
MA1-7265 has been successfully used in Western blot and Immunofluorescence applications.
The MA1-7265 antibody was raised against the Herpes simplex type 2 envelope protein.
Members of the herpes virus family (Herpesviridae), Herpes simplex virus types 1 and 2 (HSV-1 and HSV-2), are ubiquitously contagious to humans. Also known as Human herpes virus 1 and 2 (HHV-1 and -2), respectively, HSV-1 strains produce cold sores, while HSV-2 produce most genital herpes. They can be spread when an infected person is producing and shedding the virus. The virion form of herpes viruses consists of a relatively large double-stranded, linear DNA genome encased within the capsid and wrapped in the viral envelope. The different HSV strains carry over 70 open reading frames (ORFs). HSV has been linked to Alzheimer's disease.