Search Thermo Fisher Scientific
PA1-75163 specifically detects Hexestrol.
PA1-75163 has been successfully used in ELISA applications.
The PA1-75163 immunogen is Hexestrol(p-OH)-HSA.
Hexestrol is a synthetic estrogen that has been used as a hormonal antineoplastic agent.