Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
MA5-17772 targets I-Ak in FACS applications and shows reactivity with Mouse samples.
The MA5-17772 immunogen is a.TL spleen.
MHC Class II (I-Ak) serves as a critical component of the immune system, facilitating the recognition and response to extracellular pathogens and foreign substances. Its primary role is to present peptides derived from extracellular proteins to T cells for immune recognition. MHC Class II (I-Ak) is composed of an alpha and beta chain, both of which contribute to the peptide-binding groove. The location of MHC Class II (I-Ak) is restricted to antigen-presenting cells, such as dendritic cells, macrophages, and B cells. These cells are responsible for capturing and processing antigens from the extracellular environment. MHC Class II (I-Ak) consists of an extracellular domain, a transmembrane domain, and a cytoplasmic tail. The extracellular domain is responsible for interacting with the antigenic peptides, while the transmembrane domain anchors the MHC Class II molecule to the cell membrane. The cytoplasmic tail plays a role in intracellular signaling events.
HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: A alpha; H-2 class II histocompatibility antigen, A-B alpha chain; H-2 class II histocompatibility antigen, A-F alpha chain; H-2 class II histocompatibility antigen, A-K alpha chain; H-2 class II histocompatibility antigen, A-Q alpha chain; H-2 class II histocompatibility antigen, A-R alpha chain; H-2 class II histocompatibility antigen, A-S alpha chain; H-2 class II histocompatibility antigen, A-U alpha chain; histocompatibility 2, class II antigen A, alpha; IAalpha; major histocompatibility protein class II alpha chain; MHC class II antigen A alpha isoform-004; MHC-II I-A(d) alpha
Gene Aliases: Aalpha; H-2Aa; H2-Aa; H2Aa; I-Aalpha; Ia-1; Ia1; IAalpha
UniProt ID: (Mouse) P14434
Entrez Gene ID: (Mouse) 14960
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support