Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody is predicted to react with rat and mouse based on 88% sequence homology.
Suggested positive control: antigen standard for CYR61 (transient overexpression lysate), MDA-MB-231 breast cancer cell lysate.
CYR61 is a secreted heparin binding protein, encoded by a growth factor inducible immediate-early gene that associates with the extracellular matrix and connective tissue. CYR61 is a member of a distinct family of angiogenic and vasculogenic regulators designated CCN proteins, which includes connective tissue growth factor (CTGF) and the mouse CYR61 homolog, Fisp12. As an angiogenic inducer, CYR61 binds to the cell surface receptor integrin alphavbeta3, where it then stimulates cell adhesion and migration and promotes DNA synthesis of human vascular endothelial cells. Expression of CYR61 is elevated during vessel growth, wound healing and chondrocyte differentiation. CYR61 is also detected in a wide variety of tumors as it induces tumor growth and functions as a marker of tumor progression.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 3CH61; CCN family member 1; Cellular communication network factor 1; cysteine rich angiogenic inducer 61; Cysteine-rich angiogenic inducer 61; cysteine-rich heparin-binding protein 61; cysteine-rich, angiogenic inducer, 61; cysteine-rich, anigogenic inducer, 61; IBP-10; IGF-binding protein 10; IGFBP-10; insulin-like growth factor binding protein 10; Insulin-like growth factor-binding protein 10; intermediate early; Protein CYR61; Protein GIG1
Gene Aliases: AI325051; CCN1; CYR61; GIG1; IGFBP10
UniProt ID: (Human) O00622, (Mouse) P18406
Entrez Gene ID: (Rabbit) 100339657, (Human) 3491, (Mouse) 16007
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support